Learn More
- Home
- Products
- Facility Safety and Maintenance
- Labels and Label Makers
- Custom Labels
- Brady™ ToughStripe™ Floor Marking Tape
Brady™ ToughStripe™ Floor Marking Tape
Product Code. 11781685
Change view
Click to view available options
Color:
Black
Black, White
Black, Yellow
Blue
Green
Orange
Red
White
White, Red
Yellow
Width (English):
2 in.
3 in.
4 in.
Width (Metric):
5.08 cm
7.6 cm
7.62 cm
10.2 cm
Unit Size:
1 roll
| Product Code. | Color | Width (English) | Width (Metric) | unitSize |
|---|---|---|---|---|
| 11781685 | Black, White | 3 in. | 7.62 cm | 1 roll |
| 11791665 | Black | 2 in. | 5.08 cm | 1 roll |
| 11791675 | Black | 3 in. | 7.62 cm | 1 roll |
| 11791685 | Black | 4 in. | 10.2 cm | 1 roll |
| 11781675 | Black, White | 2 in. | 5.08 cm | 1 roll |
| 11781695 | Black, White | 4 in. | 10.2 cm | 1 roll |
| 11761675 | Black, Yellow | 2 in. | 5.08 cm | 1 roll |
| 11761685 | Black, White | 3 in. | 7.6 cm | 1 roll |
| 11761695 | Black, Yellow | 4 in. | 10.2 cm | 1 roll |
| 11731675 | Blue | 2 in. | 5.08 cm | 1 roll |
| 11731685 | Blue | 3 in. | 7.62 cm | 1 roll |
| 11731695 | Blue | 4 in. | 10.2 cm | 1 roll |
| 11741675 | Green | 2 in. | 5.08 cm | 1 roll |
| 11741685 | Green | 3 in. | 7.62 cm | 1 roll |
| 11741695 | Green | 4 in. | 10.2 cm | 1 roll |
| 11751675 | Orange | 2 in. | 5.08 cm | 1 roll |
| 11751685 | Orange | 3 in. | 7.62 cm | 1 roll |
| 11751695 | Orange | 4 in. | 10.2 cm | 1 roll |
| 11721675 | Red | 2 in. | 5.08 cm | 1 roll |
| 11721685 | Red | 3 in. | 7.62 cm | 1 roll |
| 11721695 | Red | 4 in. | 10.2 cm | 1 roll |
| 11771675 | White, Red | 2 in. | 5.08 cm | 1 roll |
| 11771685 | White, Red | 3 in. | 7.62 cm | 1 roll |
| 11771695 | White, Red | 4 in. | 10.2 cm | 1 roll |
| 11701675 | White | 2 in. | 5.08 cm | 1 roll |
| 11701685 | White | 3 in. | 7.62 cm | 1 roll |
| 11701695 | White | 4 in. | 10.2 cm | 1 roll |
| 11711675 | Yellow | 2 in. | 5.08 cm | 1 roll |
| 11711685 | Yellow | 3 in. | 7.62 cm | 1 roll |
| 11711695 | Yellow | 4 in. | 10.2 cm | 1 roll |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Visible floor-marking improves safety and organization in almost any industrial environment
- High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
- Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
- Can be easily applied by one person working alone
Specifications
Specifications
| Certifications/Compliance | ASTM ANSI F1679, ANSI A1264.2 2006, OSHA 1910.22 |
| Description | ToughStripe FLOOR TAPES |
| Adhesive Type | Rubber, Permanent |
| Coating | None |
| Color | Black, White |
| Finish | Gloss |
| Format | Tape |
| Material | Polyester |
| Printable Sides | Single-side Printable |
| Print Technology | None |
| Product Type | Label Tape |
| Sterility | Non-sterile |
| For Use With (Application) | Used to mark aisles, passageways, storage locations and point out safety equipment and exits |
| Gloss | 115 Gardner Units |
| Min. Operating Temperature (Metric) | -18°C |
| No. per Pack | 1 Roll/Pack |
| Quantity | 1 Roll |
| Shape | Rectangle |
| Temperature Range (English) | 0°F to 130°F |
| Temperature Range (Metric) | -18°C to +54°C |
| Thickness (English) | 0.008 in. |
| Thickness (Metric) | 0.201 mm |
| Dimensions (LxW) | 100 ft. x 3 in. (30.48 m x 7.62 cm) |
| Length (English) | 100 ft. |
| Length (Metric) | 30.48 m |
| Width (English) | 3 in. |
| Width (Metric) | 7.62 cm |
| Alternate Part Number | TS-76.20-514-BK/WT-STR-RL |
| Show More Show Less |