Learn More
Brady™ ToughStripe™ Floor Marking Center Marks
Product Code. 11731246
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
Length (English):
5 in.
8 in.
10 in.
Length (Metric):
12.7 cm
20.32 cm
25.4 cm
Width (English):
2 in.
3 in.
4 in.
Unit Size:
1 pack
Product Code. Color Length (English) Length (Metric) Width (English) unitSize
11731246 White 5 in. 12.7 cm 2 in. 1 pack
11701246 Blue 5 in. 12.7 cm 2 in. 1 pack
11711246 Yellow 5 in. 12.7 cm 2 in. 1 pack
11721246 Black 5 in. 12.7 cm 2 in. 1 pack
11781236 Red 5 in. 12.7 cm 2 in. 1 pack
11791236 Green 5 in. 12.7 cm 2 in. 1 pack
11751246 Green 8 in. 20.32 cm 3 in. 1 pack
11741246 Red 8 in. 20.32 cm 3 in. 1 pack
11771246 Yellow 8 in. 20.32 cm 3 in. 1 pack
11791246 White 8 in. 20.32 cm 3 in. 1 pack
11781246 Black 8 in. 20.32 cm 3 in. 1 pack
11701256 Red 10 in. 25.4 cm 4 in. 1 pack
11761246 Blue 8 in. 20.32 cm 3 in. 1 pack
11711256 Green 10 in. 25.4 cm 4 in. 1 pack
11721256 Blue 10 in. 25.4 cm 4 in. 1 pack
11751256 White 10 in. 25.4 cm 4 in. 1 pack
11731256 Yellow 10 in. 25.4 cm 4 in. 1 pack
11741256 Black 10 in. 25.4 cm 4 in. 1 pack
18 options
Product Code. 11731246
Brand: Brady™ 104471
Only null left
Add to basket
Add to basket
ToughStripe™ Floor Marking Center Marks are extremely durable die-cut floor marking shapes made of rugged polyester with ultra-aggressive adhesive and hold up against forklift traffic.
  • Visible floor-marking improves safety and organization in almost any industrial environment
  • High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
  • Ideal for marking corners of work cells and storage locations
  • Can be easily applied by one person working alone
Specifications
Certifications/Compliance ASTM ANSI F1679, ANSI A1264.2 2006, OSHA 1910.22
Description ToughStripe Diecut Shapes (L's, T's, +'s, footprints and more)
Adhesive Type Rubber, Permanent
Coating None
Color White
Finish Gloss
Format Tape
Material Polyester
Printable Sides Single-side Printable
Print Technology None
Product Type Label Tape
Sterility Non-sterile
For Use With (Application) For floor corner mark, Used to mark aisles, passageways, storage locations and point out safety equipment and exits
Gloss 115 Gardner Units
Min. Operating Temperature (Metric) -18°C
No. per Pack 20 Pieces/Pack
Quantity 20/Pk.
Shape Cross
Temperature Range (English) 0°F to 130°F
Temperature Range (Metric) -18°C to +54°C
Thickness (English) 0.008 in.
Thickness (Metric) 0.201 mm
Dimensions (LxW) 127.00 x 50.8 mm
Length (English) 5 in.
Length (Metric) 12.7 cm
Width (English) 2 in.
Width (Metric) 50.8 mm
Height (English) 5 in.
Height (Metric) 127.00 mm
Alternate Part Number TS-50.80-514-CEN-WT-20