Learn More
Brady™ TLS2200™ and TLS-PC™ Vial, Centrifuge or Eppendorf Tubes™ Labels
Product Code. 10733183
Change view
Click to view available options
Quantity:
100/Roll
500/Pk.
Height (Metric):
12.7 mm
25 mm
55.88 mm
66.675 mm
Material:
Nylon Cloth
Polyester
Vinyl
Unit Size:
1 roll
Product Code. Quantity Height (Metric) Material unitSize
10733183 100/Roll 66.675 mm Polyester 1 roll
10512212 500/Pk. 25 mm Nylon Cloth 1 roll
10618922 100/Roll 12.7 mm Polyester 1 roll
11532905 100/Roll 55.88 mm Vinyl 1 roll
4 options
Product Code. 10733183
Brand: Brady™ 062278
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
TLS2200™ and TLS-PC™ Vial, Centrifuge or Eppendorf Tubes™ Labels are ideal for cables, laboratory ID or general labeling where graphics need to be protected.
  • Labels for the TLS 2200™ and TLS-PC Link™ thermal transfer Portable Printers.
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.
  • Matte white polyamide coated nylon cloth material (B-499) has a permanent acrylic adhesive with a service temperature of -40 to 193° F (-40 to 90° C) and recommended for indoor use.
  • Self-Laminating Polyester (B-461) has a permanent acrylic adhesive , good print smudge resistance, solvent resistance and high/low temperature performance.
Specifications
Certifications/Compliance RoHS
Description TLS 2200/TLS PC Link Labels
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Format Label
Material Polyester
Product Type TLS 2200™/TLS-PC Link™ Label
Sterility Non-sterile
For Use With (Application) Centrifuge tube, Cryo vial, Eppendorf tube, Laboratory identification, PCR tube
For Use With (Equipment) BMP™61 Labeling System; BMP™71 Label Printer; TLS 2200™ Thermal Labeling System; TLS-PC LINK™ Thermal Labeling System
Labels per Row 1
Quantity 100/Roll
Shape Rectangle
Temperature Range (Metric) -196 °C to 110 °C
Thickness (English) 0.0019 in.
Thickness (Metric) 0.048 mm
Vertical Repeat (Metric) 69.85 mm
Dimensions (LxW) 2.625 in. H x 0.6 in. W
Width (English) 0.6 in.
Width (Metric) 15.24 mm
Height (English) 2.62 in.
Height (Metric) 66.675 mm