Learn More
Brady™ TLS 2200™/TLS PC Link™ Portable Thermal Transfer Printer Labels for Vial Tops
Product Code. 10763293
Change view
Click to view available options
Color:
White
Yellow
Shape:
Rectangle with Rounded Corners
Round
Diameter (English):
0.3 in.
0.375 in.
0.5 in.
Unit Size:
1 roll
Product Code. Color Shape Diameter (English) unitSize
10763293 Yellow Round 0.375 in. 1 roll
10151342 Yellow Round 0.5 in. 1 roll
10742223 White Rectangle with Rounded Corners 0.3 in. 1 roll
12877258 White Round 0.375 in. 1 roll
10520183 White Round 0.5 in. 1 roll
5 options
Product Code. 10763293
Brand: Brady™ 062252
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Labels for vial tops
  • Durable, chemical- and solvent-resistant labels; ink will not smear or fade, and the text stays crisp and readable
  • Printable labels allow more information to fit on any size legend
  • Choice of materials suitable for different environments from liquid nitrogen to autoclaves
Order Info
Labels available in other materials and sizes; contact your Customer Service Representative for ordering information.
Specifications
Certifications/Compliance Halogen-free (DIN VDE 0472/parts 815), UL
Description Surface Mount
Adhesive Type Acrylic, Permanent
Color Yellow
Finish Matte
Format Label
Material Nylon Cloth
Product Type TLS 2200™/TLS-PC Link™ Label
For Use With (Application) Laboratory identification, vial, vial top
For Use With (Equipment) BMP™71, TLS 2200™, TLS-PC Link™
Labels per Row 1
Quantity 500/Pk.
Shape Round
Temperature Range (English) -320-194°F
Temperature Range (Metric) -196°C to +90°C
Thickness (Metric) 0.165 mm
Vertical Repeat (Metric) 12.7 mm
Diameter (English) 0.375 in.
Diameter (Metric) 9.5 mm