Learn More
Brady™ Nylon Cloth Laboratory Labels
Product Code. 13533310
Change view
Click to view available options
Width (English):
1 in.
2 in.
Height (English):
0.25 in.
0.3 in.
Height (Metric):
6 mm
9 mm
Unit Size:
Each
Pack of 1500
Product Code. Width (English) Height (English) Height (Metric) unitSize
13533310 1 in. 0.3 in. 9 mm Each
15706879 2 in. 0.25 in. 6 mm Pack of 1500
2 options
Product Code. 13533310
Brand: Brady™ 143017
Only null left
Add to basket
Add to basket
Nylon Cloth Laboratory Labels (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
  • Matte white polyamide coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable
  • Recommended for indoor use
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
  • Service temperature of -40 to 193° F (-40 to 90° C)
Specifications
Autoclavable Autoclavable
Certifications/Compliance RoHS
Description Nylon Cloth Labels for the BBP33 Printer
Adhesive Type Acrylic, Permanent
Color White
Display Consignment
Finish Matte
Format Die-cut
Material Nylon Cloth
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Laboratory Label
Sterility Non-sterile
For Use With (Application) Wiremarking and general labeling applications
For Use With (Equipment) BBP™ 33
Horizontal Repeat (Metric) 27.94 mm
Labels per Row 2
Min. Operating Temperature (Metric) -40°C
No. per Pack 1500 Labels/Box
Quantity 1500/Pk.
Shape Rectangle
Software Type LabelMark, MarkWare
Temperature Range (English) -40°F to 193°F
Temperature Range (Metric) -40°C to +90°C
Tensile Strength 80 lb./in.
Thickness (English) 0.0065 in.
Thickness (Metric) 0.165 mm
Vertical Repeat (Metric) 12.07 mm
Dimensions (LxW) 0.375 x 1 in.
Length (English) 0.3 in.
Width (English) 1 in.
Width (Metric) 25.4 mm
Width (Metric) Printable Area 58.42 mm
Height (English) 0.3 in.
Height (Metric) 9 mm
Alternate Part Number B33-152-499