Learn More
Brady™ Labels for Desktop Laser Printers
Labels for Desktop Laser Printers ideal for rating plates, asset identification, and general purpose labeling applications.
Brand: Brady™ 089405
1195.81 NOK valid until 2026-10-31
BEST PRICE on Promo! Use promo code "28008" to get your promotional price.
Description
- Durable, chemical- and solvent-resistant labels for use with a variety of laser printers.
- High resolution, good solvent and smudge resistance, moderate to high temperature performance.
- Withstand extreme laboratory environments.
Specifications
| Autoclavable | |
| LaserTab Laser Printable Labels | |
| White | |
| Matte | |
| Nylon Cloth | |
| Laser | |
| Non-sterile | |
| Laser Printers | |
| Pack of 9 Sheets | |
| -40°C | |
| 2500 Pk. | |
| 0.375 in. dia. | |
| -40°C to +90°C | |
| 0.165 mm | |
| 0.375 in. | |
| 9.52 mm | |
| 0.37 in. | |
| LAT-60-799-2.5 |
| Halogen Free (DIN VDE 0472/part 815) | |
| Acrylic, Permanent | |
| Consignment | |
| Label | |
| Single-side Printable | |
| Label | |
| Freezers, Liquid Nitrogen, Autoclaves, Hot Water Baths, Laboratory Labeling and Identification | |
| 12.70 mm | |
| 15 | |
| 2565 Labels/Pack | |
| Round | |
| -40°F to +194°F | |
| 6.5 mil | |
| 0.375 x 0.375 in. (9.35 x 9.35 mm) | |
| 0.375 in. | |
| 12.70 mm | |
| 0.952 cm |