Learn More
Brady™ Label Maker Vinyl Cartridges (B-595) for BMP51, BMP53, BMP41 Printers
Description
- Durable and low-shrink with aggressive adhesive
- Used on surfaces such as pipes, walls, doors, panels, shelving, bins and equipment
- Offers resistance to oils, detergents, dirt, grime and many solvents
Specifications
Specifications
| Color | Black, Yellow |
| Material | Vinyl (B-595) |
| Product Type | Label Maker Cartridge |
| For Use With (Equipment) | BMP™51,BMP™53,BMP™41 Printers |
| Quantity | 1 Ea. |
| Size (English) | 25 ft. x 1 in. |
| Size (Metric) | 7.62 m x 25.4 mm |
| Length (English) | 25 ft. |
| Width (English) | 1 in. |
| Width (Metric) | 25.4 mm |
| Show More |