Learn More
Brady™ Label Maker Vinyl Cartridges (B-427) for BMP51, BMP53, BMP41 Printers
Product Code. 15123339
Change view
Click to view available options
Color:
Black, White, Clear
Black, Yellow
Size (Metric):
12.7 x 19.050 mm
12.7 x 25.4 mm
12.7 x 31.75 mm
12.7 x 38.1 mm
19.050 x 25.4 mm
19.050 x 38.1 mm
25.4 x 101.6 mm
25.4 x 19.050 mm
25.4 x 25.4 mm
25.4 x 31.750 mm
25.4 x 38.1 mm
25.4 x 63.5 mm
Size (English):
0.5 x 0.75 in.
0.5 x 1 in.
0.5 x 1.25 in.
0.5 x 1.5 in.
0.75 x 1 in.
0.75 x 1.5 in.
1 x 0.75 in.
1 x 1 in.
1 x 1.25 in.
1 x 1.5 in.
1 x 2.5 in.
1 x 4 in.
Unit Size:
Each
Product Code. Color Size (Metric) Size (English) unitSize
15123339 Black, Yellow 25.4 x 63.5 mm 1 x 2.5 in. Each
15103339 Black, White, Clear 12.7 x 38.1 mm 0.5 x 1.5 in. Each
15153339 Black, Yellow 25.4 x 38.1 mm 1 x 1.5 in. Each
15133339 Black, White, Clear 25.4 x 38.1 mm 1 x 1.5 in. Each
15113339 Black, White, Clear 19.050 x 38.1 mm 0.75 x 1.5 in. Each
15143339 Black, Yellow 25.4 x 101.6 mm 1 x 4 in. Each
15163329 Black, White, Clear 25.4 x 63.5 mm 1 x 2.5 in. Each
15133329 Black, White, Clear 19.050 x 25.4 mm 0.75 x 1 in. Each
15173329 Black, White, Clear 25.4 x 101.6 mm 1 x 4 in. Each
15153329 Black, White, Clear 25.4 x 25.4 mm 1 x 1 in. Each
15143329 Black, White, Clear 12.7 x 25.4 mm 0.5 x 1 in. Each
15193329 Black, White, Clear 25.4 x 31.750 mm 1 x 1.25 in. Each
15123329 Black, White, Clear 25.4 x 19.050 mm 1 x 0.75 in. Each
11942782 Black, White, Clear 12.7 x 31.75 mm 0.5 x 1.25 in. Each
11972772 Black, White, Clear 12.7 x 19.050 mm 0.5 x 0.75 in. Each
15 options
Product Code. 15123339
Brand: Brady™ 131585
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed to adhere to highly curved surfaces, wires and cables
  • Feature printable zone in white or yellow
  • Transparent, non printable area wraps around wires and cables for laminating printed texts
  • Offers resistance to abrasion, handling, solvents, water, oil and dirt
  • Available in various sizes
Specifications
Color Black, Yellow
Material Self-laminating Vinyl (B-427)
Product Type Label Maker Cartridge
For Use With (Equipment) BMP™51,BMP™53,BMP™41 Printers
Quantity 1 Ea. (110 labels)
Size (English) 1 x 2.5 in.
Size (Metric) 25.4 x 63.5 mm
Width (English) 1 in.
Width (Metric) 25.4 mm
Height (English) 2.5 in.
Height (Metric) 63.5 mm