Learn More
Brady™ Label Maker Cartridge: Polyester, StainerBondz™
Click to view available options
Height (English):
0.3 in.
0.75 in.
0.9 in.
Unit Size:
Each
Description
- StainerBondz™ B-481 matte white polyester has a permanent acrylic adhesive and is thermal transfer printable with B30 series printers from Brady™
- B-481 performs well in the commonly used H & E (hematoxilin and eosin) slide staining process
- Excellent resistance to a wide variety of chemicals, oils and water
- All-in-one cartridge includes label supply and R6400 Series ribbon
Specifications
Specifications
| Certifications/Compliance | RoHS, ASTM D |
| Adhesive Type | Acrylic |
| Color | Black |
| Finish | Matte White |
| Material | Polyester |
| Sterility | Non-sterile |
| For Use With (Application) | Slide staining process and other laboratory identification which requires superior chemical resistance |
| For Use With (Equipment) | BMP53 Label Printer |
| Quantity | 1 Ea. |
| Temperature Range (English) | -112 to 266°F |
| Show More |