Learn More
Brady™ FreezerBondz II Polyester Laboratory Labels B490
Product Code. 17878363
Change view
Click to view available options
Height (English):
0.5 in.
0.6 in.
1.25 in.
Quantity:
1500/Pk.
1650/Pk.
Unit Size:
1 roll
Each
Width (English):
0.3 in.
1.6 in.
Product Code. Height (English) Quantity unitSize Width (English)
17878363 0.5 in. 1650/Pk. 1 roll -
15868821 0.6 in. 1500/Pk. 1 roll 1.6 in.
15313744 1.25 in. 1500/Pk. Each 0.3 in.
3 options
Product Code. 17878363
Brand: Brady™ 142938
FreezerBondz II Matte White Polyester Laboratory Labels are durable polyester markers designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
  • B-490 labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • White polyester material has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance of -320 to 212° F (-196 to 100° C)
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • Brady Series R4300 ribbon is recommended
Specifications
Certifications/Compliance RoHS, 2002/618/EC MCV, 2002895/EC
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Format Die-cut
Material Polyester
Print Technology Thermal Transfer
For Use With (Application) For use in laboratory identification such as vials, centrifuge tubes, test tubes, straws and slides
For Use With (Equipment) BBP™ 33 Printer
Labels per Row 1
Min. Operating Temperature (Metric) -196°C
No. per Pack 1650 Labels/Box
Quantity 1650/Pk.
Shape Rectangle
Temperature Range (English) -320°F to +266°F
Temperature Range (Metric) -196°C to +130°C
Thickness (Metric) 0.065 mm
Length (English) 0.5 in.
Height (English) 0.5 in.
Height (Metric) 12.70 mm