Learn More
Brady™ BMP™ 21 ID Pal™ Label Cartridges: Self Laminating Vinyl
Product Code. p-3898617
Click to view available options
Width (English):
0.75 in.
1 in.
1.5 in.
Unit Size:
Each
Product Code. 11543894
Brand: Brady™ 110928
Only null left
Add to basket
Add to basket
Self-laminating vinyl thermal transfer labels.
  • Sticks well to both smooth and textured surfaces.
  • Allows you to label cables and wires with a simple wrapping function.
  • Provides excellent smudge resistant text.
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance UL
Description Self-laminating Vinyl tape for M210, M210-LAB, M211, BMP21-PLUS, BMP21-LAB
Adhesive Type Permanent Acrylic
Color White, Translucent, Black
Finish Matte
Format Cartridge (Label)
Material Vinyl
Product Type Label
Sterility Non-sterile
For Use With (Application) M210, M210-LAB, M211, BMP21-PLUS, BMP21-LAB, IDPAL, LABPAL
For Use With (Equipment) M210;M210-LAB;M211;BMP21-PLUS;BMP21-LAB;IDPAL;LABPAL
Min. Operating Temperature (Metric) -70°C
Quantity 1 Roll
Shape Rectangle
Temperature Range (English) -94°F to +158°F
Temperature Range (Metric) -70°C to +70°C
Thickness (English) 0.004 in
Thickness (Metric) 0.102 mm
Wire Gauge 16 to 8 AWG
Dimensions (LxW) 25.40 mm (W) x 4.30 m (L)
Length (English) 14 ft.
Length (Metric) 4.30 m
Width (English) 1 in.
Width (Metric) 25.40 mm
Width (Metric) Printable Area 9.53 mm
Width (Metric) Tape 25.40 mm
Height (English) 14 ft.
Height (Metric) 4.2 m
Alternate Part Number M21-1000-427