Learn More
Brady™ BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL, Matte White Tape
GREENER_CHOICE
Product Code. p-4909579
Change view
Click to view available options
Width (English):
0.3 in.
0.5 in.
0.75 in.
Unit Size:
Each
Product Code. Width (English) unitSize
12837268 0.3 in. Each
11583894 0.5 in. Each
11593894 0.75 in. Each
3 options
Product Code. 12837268
Brand: Brady™ 110931
Edge
Use this film tape for a variety of labeling applications. BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL are ideal for centrifuge, Eppendorf and PCR tubes; conical and large tubes; cryo tubes and vials.
  • High adhesive
  • Translucent
  • Gloss finish
  • Perfect for autoclaving and for use in below freezing temperatures
Specifications
Certifications/Compliance Halogen-free (DIN VDE 0472/parts 815)
Adhesive Type Permanent Acrylic
Color Black, White
Finish Matte
Format Cartridge (Label)
Material Polyester
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) General Identification, Laboratory Identification: Vial, Well Plate
For Use With (Equipment) M210 Label Printer, M210-LAB Label Printer, M211 Label Printer, BMP™ 21 LAB Label Printer, BMP™ 21-PLUS Label Printer
Quantity 1 Roll
Dimensions (LxW) 6.40 m x 9.53 mm
Length (English) 21 ft.
Width (English) 0.3 in.
Width (Metric) 9.53 mm
Height (English) 21 ft.
Height (Metric) 6.4 cm
Alternate Part Number M21-375-461-AW