Learn More
Brady™ BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL, Matte White Tape
Product Code. 11583894 Shop All Brady Products
Change view
Click to view available options
Width (English):
0.3 in.
0.5 in.
0.75 in.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Unit Size
11583894 0.5 in. Each
12837268 0.3 in. Each
11593894 0.75 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11583894 Supplier Brady™ Supplier No. 110932
  • ONLINE EXCLUSIVE PRICE
    627.00 NOK
    554.47 NOK / Pack of 1
    Save 72.53 NOK
    11% Off
In Stock
Add to basket
Edge
Add to basket
Use this film tape for a variety of labeling applications. BMP™ Label Cartridges for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL are ideal for centrifuge, Eppendorf and PCR tubes; conical and large tubes; cryo tubes and vials.
  • High adhesive
  • Translucent
  • Gloss finish
  • Perfect for autoclaving and for use in below freezing temperatures
Specifications
Certifications/Compliance Halogen-free (DIN VDE 0472/parts 815)
Description Polyester tape for BMP21-PLUS; BMP21-LAB; BMP21; LABPAL
Adhesive Type Permanent Acrylic
Color Black, White
Finish Matte
Format Cartridge (Label)
Material Polyester
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) General Identification, Laboratory Identification: Vial, Well Plate
For Use With (Equipment) M210 Label Printer, M210-LAB Label Printer, M211 Label Printer, BMP™ 21 LAB Label Printer, BMP™ 21-PLUS Label Printer
Labels per Row 1
Quantity 1 Roll
Temperature Range (Metric) -196°C to +110°C
Thickness (Metric) 0.048 mm
Dimensions (L x W) 6.40 m x 12.7 mm
Length (English) 21 ft.
Length (Metric) 6.40 m
Width (English) 0.5 in.
Width (Metric) 12.70 mm
Height (English) 21 ft.
Height (Metric) 6.4 cm
Alternate Part Number M21-500-461-AW