Learn More
Brady™ BBP™ Hi-Performance Polyester Tape
Click to view available options
Color:
Black
Blue
Clear
Green
Orange
Red
White
Yellow
Width (English):
0.5 in.
1.12 in.
1.125 in.
2.25 in.
3 in.
4 in.
Unit Size:
Each
Description
- Durable, ultra-smooth polyester labels with permanent pressure sensitive acrylic adhesive
- Outdoor grade polyester tape can be thermal transfer printed
- Withstands mild chemicals, oil and water
- Enables printers with XY cutters to cut custom shapes or multiple labels out of a single continuous roll
Specifications
Specifications
| Description | High Performance Polyester Tape for BBP3x Printers |
| Adhesive Type | Acrylic, Permanent, Pressure-sensitive |
| Color | Red |
| Finish | Gloss |
| Format | Tape |
| Material | Polyester |
| Sterility | Non-sterile |
| For Use With (Application) | Designed for use on Stainless Steel pipes in harsh environments such as the pulp and Paper industry |
| For Use With (Equipment) | BBP30, BBP31, BBP33, BBP35, and BBP37 printers |
| Gloss | 80 Gardner Units |
| Show More |