Learn More
Brady™ BBP™ Blank White Pre-Cut Blank Sign Headers
Click to view available options
Dimensions (LxW):
2.9 H x 7.2 cm W
4 x 6 in.
5.7 H x 7.6 cm W
5.7 H x 9.8 cm W
Quantity:
175/Pk.
250/Pk.
300/Pk.
Unit Size:
Each
Description
- Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces—including outdoors
- Conforms and adheres to "difficult" surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
- Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
- 8-10 year average outdoor durability in temperatures from -40°F to 180°F (-40°C to 82°C); can be applied as low as 0°F (-18°C)
Specifications
Specifications
| Certifications/Compliance | ASME ANSI Z535, OSHA 1910.145 |
| Description | Outdoor Grade Vinyl Labels for BBP3x Printers |
| Adhesive Type | Permanent Acrylic |
| Color | White |
| Finish | Gloss |
| Material | Vinyl |
| Product Type | Pre-cut Blank Label |
| For Use With (Application) | Pipemarkers, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels |
| For Use With (Equipment) | BBP30, BBP31, BBP33, BBP35, and BBP37 printers |
| Quantity | 250/Pk. |
| Show More |