Learn More
Brady™ B-427 Self Laminating Vinyl Bulk BMP71 Printer Labels
Product Code. 11791755
Change view
Click to view available options
Dimensions (LxW):
38.10 x 25.40 mm
63.50 x 25.40 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxW) unitSize
11791755 63.50 x 25.40 mm Each
11781805 38.10 x 25.40 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11791755 Supplier Brady™ Supplier No. 114873
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
White/translucent label for wire and cable marking
  • Features white printable area with clear “tail” that wraps around marker
  • Resistant to abrasion, solvents, water, oil and dirt
Applications:
Data Communications, Electrical, Laboratory, Healthcare, Cable Labels
Certifications
UL listed
Specifications
Certifications/Compliance UL Recognised to UL969 Labelling and Marking Standard when printed with the Brady Series R4300 and R6200 ribbons. See UL files MH17154 for specific details. Brady customers should specify at purchase if they require UL Recognised B-427 labels. Brady sel
Description Bulk BMP71 Label Printer Labels
Adhesive Type Permanent Acrylic
Color White, Translucent
Finish Matte
Material Self-laminating Vinyl (B-427)
Product Type Bulk Labels
For Use With (Application) Communications cables, Laboratory identification, Straws, Voice/data identification
For Use With (Equipment) BMP™71 Printer
Labels per Row 1
Quantity 1000/Roll
Shape Rectangle
Size (English) 1 x 2.5 in.
Size (Metric) 25.4 x 25.4 mm
Temperature Range (Metric) -70°C to +70°C
Thickness (Metric) 0.102 mm
Vertical Repeat (Metric) 66.68 mm
Wire Gauge 3 to 0000 AWG
Dimensions (LxW) 63.50 x 25.40 mm
Width (English) 1 in.
Width (Metric) 25.4 mm
Height (English) 2.5 in.
Height (Metric) 63.5 mm
Height (Metric) Printable Area 19.05 mm