Learn More
Trajan™ BPX5 GC Capillary Columns
Product Code. 10430081
Click to view available options
Length (Metric):
10 m
12 m
15 m
25 m
30 m
50 m
6 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.15 μm
0.25 μm
0.5 μm
1 μm
Unit Size:
Each
Product Code. 10430081
Brand: Trajan™ 054125
Only null left
Add to basket
Add to basket
MS premium, low bleed phase designed for robustness and is suitable for a wide range of applications
  • Very low bleed
  • High temperature
  • Non-polar
  • Ideal for GCMS
  • Applications include pesticides, herbicides, hydrocarbons, amines and phenols
Specifications
Max. Temperature 370°C
Functionality/Interaction Type Pi Bonding with the Aromatic Phenyl Groups
Diameter (Metric) Inner 0.32 mm
Stationary Phase 5% Phenyl Polysilphenylene Siloxane
USP Type G27, G36, G41
For Use With (Application) Amines Analysis, Pesticides and Herbicides Analysis, Hydrocarbons, Solvents Analysis, Phenols, Amines, GC/MS
Temperature Range -40°C to +360/370°C
Film Thickness 0.5 μm
Length (Metric) 25 m
Packing Material BPX5
Polarity Non polar
Coating Fused Silica