Learn More
Trajan Scientific and Medical™ BPX5 GC Capillary Columns
Product Code. 15228653
Click to view available options
Length (Metric):
30 m
60 m
Diameter (Metric):
0.25 mm
0.32 mm
Film Thickness:
0.5 μm
1 μm
Unit Size:
Each
Product Code. 15228653
Brand: Trajan Scientific and Medical™ 054153
Only null left
Add to basket
Add to basket
Suitable for a wide range of applications. Trajan Scientific and Medical BPX5 GC Capillary Columns are MS premium, low bleed phase designed for robustness.
  • Very low bleed
  • High temperature
  • Non-polar
  • Ideal for GCMS
  • Applications include pesticides, herbicides, hydrocarbons, amines and phenols
Specifications
Diameter (Metric) 0.32 mm
Type BPX5 GC Capillary Column
Diameter (Metric) Inner 0.32 mm
For Use With (Application) Ultra Trace Analyses Analysis, Pesticides and Herbicides Analysis, Hydrocarbons, Solvents Analysis, Phenols, Amines, GC-MS and Other Specific Detector Applications
Temperature Range -40°C to +360/370°C
Film Thickness 1 μm
Length (Metric) 30 m
Material 5% Phenyl Polysilphenylene-siloxane
Packing Material BPX5