Learn More
Trajan Scientific and Medical™ BPX5 GC Capillary Columns
Click to view available options
Length (Metric):
30 m
60 m
Diameter (Metric):
0.25 mm
0.32 mm
Film Thickness:
0.5 μm
1 μm
Unit Size:
Each
Description
- Very low bleed
- High temperature
- Non-polar
- Ideal for GCMS
- Applications include pesticides, herbicides, hydrocarbons, amines and phenols
Specifications
Specifications
| Diameter (Metric) | 0.25 mm |
| Type | BPX5 GC Capillary Column |
| For Use With (Application) | Ultra Trace Analyses Analysis, Pesticides and Herbicides Analysis, Hydrocarbons, Solvents Analysis, Phenols, Amines, GC-MS and Other Specific Detector Applications |
| Temperature Range | -40°C to +360/370°C |
| Film Thickness | 1 μm |
| Length (Metric) | 60 m |
| Material | 5% Phenyl Polysilphenylene-siloxane |
| Packing Material | BPX5 |