Learn More
BPIFA2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28293-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16846085
  • 1415.13 NOK / 20µL
Estimated Shipment: 16-10-2023
Description
The BPIFA2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BPIFA2. This antibody recognizes human antigen. The BPIFA2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
BPIFA2
Polyclonal
Unconjugated
20 μL
140683
IgG
Western Blot, ELISA
367 μg/mL