Learn More
BPGM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17173-1-AP-150UL
 View more versions of this product
Product Code. 16837014
  • 4836.10 NOK / 150µL
Description
BPGM belongs to the phosphoglycerate mutase family, BPG-dependent PGAM subfamily. It plays a major role in regulating hemoglobin oxygen affinity as a consequence of controlling 2, 3-BPG concentration. It can also catalyze the reaction of EC 5.4.2.1 (mutase) and EC 3.1.3.13 (phosphatase), but with a reduced activity.
Specifications
Specifications
BPGM
Polyclonal
Unconjugated
BPGM
2,3 bisphosphoglycerate mutase, Bisphosphoglycerate mutase, BPG dependent PGAM, BPGM
Rabbit
Antigen Affinity Chromatography
RUO
12183, 296973, 669
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07738, P15327
BPGM
BPGM Fusion Protein Ag10731
150 μL
Primary
Human, Mouse, Rat