Learn More
Trajan Scientific and Medical™ BP5 GC Capillary Columns
Product Code. 15288653
Click to view available options
Diameter (Metric):
0.25 mm
0.53 mm
Unit Size:
Each
Product Code. 15288653
Brand: Trajan Scientific and Medical™ 054195
Only null left
Add to basket
Add to basket
Used for a wide range of applications. Trajan Scientific and Medical BP5 GC Capillary Columns are general purpose phase columns.
  • Excellent general purpose column
  • Low bleed
  • Non-polar High temperature
  • Ideal for aromatics, pesticides, herbicides, drugs of abuse and hydrocarbons
Specifications
Diameter (Metric) 0.53 mm
Type BP5 GC Capillary Column
For Use With (Application) Hydrocarbons Analysis, Pesticide Analysis, Herbicides Analysis, Drugs of Abuse, Hydrocarbons, GC/MS
Temperature Range -60°C to +320/340°C
Film Thickness 1 μm
Length (Metric) 30 m
Material 5% Phenyl, 95% Dimethyl Polysiloxane
Packing Material BP5