Learn More
Trajan Scientific and Medical™ BP5 GC Capillary Columns
Product Code. 15218663
Click to view available options
Diameter (Metric):
0.25 mm
0.53 mm
Unit Size:
Each
Product Code. 15218663
Brand: Trajan Scientific and Medical™ 054203
Only null left
Add to basket
Add to basket
Used for a wide range of applications. Trajan Scientific and Medical BP5 GC Capillary Columns are general purpose phase columns.
  • Excellent general purpose column
  • Low bleed
  • Non-polar High temperature
  • Ideal for aromatics, pesticides, herbicides, drugs of abuse and hydrocarbons
Specifications
Diameter (Metric) 0.25 mm
Type BP5 GC Capillary Column
For Use With (Application) Drugs of Abuse Analysis, Pesticide Analysis, Herbicides Analysis, Drugs of Abuse, Hydrocarbons, GC/MS
Temperature Range -60°C to +320/340°C
Film Thickness 1 μm
Length (Metric) 30 m
Material 5% Phenyl, 95% Dimethyl Polysiloxane
Packing Material BP5