Learn More
Trajan Scientific and Medical™ BP1 PONA GC Capillary Columns
Click to view available options
Length (Metric):
60 m
Unit Size:
Each
Description
- Classic cross-linked dimethyl polysiloxane technology
- Low bleed
- Non-polar
- Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
Specifications
| Diameter (Metric) | 0.53 mm |
| Type | BP1 PONA GC Capillary Column |
| For Use With (Application) | Petroleum Hydrocarbons Analysis, Gasoline Range Hydrocarbons, MTBE |
| Temperature Range | -60°C to +340/360°C |
| Film Thickness | 3 μm |
| Length (Metric) | 60 m |
| Material | 100% Dimethyl Polysiloxane |
| Packing Material | BP1 PONA |