Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- GC Columns
- GC Capillary Columns
- Trajan™ BP1 GC Capillary Columns
Trajan™ BP1 GC Capillary Columns
Product Code. 16602612
Change view
Click to view available options
Length (Metric):
12 m
12 mm
25 m
30 m
50 m
60 m
60 mm
Diameter (Metric) Inner:
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.5 μm
1 μm
3 μm
5 μm
0.25 mm
Unit Size:
Each
| Product Code. | Length (Metric) | Diameter (Metric) Inner | Film Thickness | unitSize |
|---|---|---|---|---|
| 16602612 | 25 m | 0.53 mm | 1 μm | Each |
| 10422171 | 25 m | 0.15 mm | 1 μm | Each |
| 10107540 | 25 m | 0.32 mm | 1 μm | Each |
| 16682602 | 12 m | 0.32 mm | 0.5 μm | Each |
| 16692602 | 50 m | 0.32 mm | 0.5 μm | Each |
| 12965531 | 50 m | 0.53 mm | 5 μm | Each |
| 12653016 | 12 m | 0.32 mm | 1 μm | Each |
| 10045400 | 25 m | 0.22 mm | 0.25 μm | Each |
| 10229050 | 50 m | 0.22 mm | 0.25 μm | Each |
| 16672602 | 60 m | 0.22 mm | 0.25 μm | Each |
| 15238633 | 30 m | 0.25 mm | 1 μm | Each |
| 10310371 | 25 m | 0.32 mm | 0.25 μm | Each |
| 15268633 | 60 m | 0.32 mm | 0.5 μm | Each |
| 10441031 | 25 m | 0.32 mm | 0.5 μm | Each |
| 15278633 | 30 m | 0.32 mm | 3 μm | Each |
| 15298633 | 60 m | 0.32 mm | 5 μm | Each |
| 10762372 | 12 m | 0.53 mm | 1 μm | Each |
| 15208713 | 30 m | 0.53 mm | 5 μm | Each |
| 15218713 | 60 m | 0.53 mm | 5 μm | Each |
| 15238713 | 60 m | 0.25 mm | 0.5 μm | Each |
| 10372160 | 60 m | 0.53 mm | 3 μm | Each |
| 12796557 | 30 m | 0.53 mm | 3 μm | Each |
| 15258713 | 30 m | 0.25 mm | 0.5 μm | Each |
| 12945531 | 60 mm | 0.25 mm | 0.25 mm | Each |
| 10298760 | 12 mm | 0.22 mm | 0.25 mm | Each |
| 10177250 | 12 m | 0.22 mm | 0.1 μm | Each |
| 12975531 | 30 m | 0.32 mm | 0.25 μm | Each |
| 12654945 | 30 m | 0.25 mm | 0.25 μm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
- Classic cross-linked dimethyl polysiloxane technology
- Low bleed
- Non-polar
- Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
Specifications
| Max. Temperature | 340/360°C |
| Functionality/Interaction Type | Van der Waals interaction |
| Diameter (Metric) Inner | 0.53 mm |
| Stationary Phase | 100% Dimethyl Polysiloxane |
| USP Type | G1, G2, G38 |
| For Use With (Application) | Amines Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines |
| Temperature Range | -60°C to +340/360°C |
| Film Thickness | 1 μm |
| Length (Metric) | 25 m |
| Packing Material | Forte™ BP1 |
| Polarity | Non polar |
| Coating | Fused Silica |
| Show More Show Less |