Learn More
Trajan™ BP1 GC Capillary Columns
Product Code. 15278633
Change view
Click to view available options
Length (Metric):
12 m
12 mm
25 m
30 m
50 m
60 m
60 mm
Diameter (Metric) Inner:
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.5 μm
1 μm
3 μm
5 μm
0.25 mm
Unit Size:
Each
Product Code. Length (Metric) Diameter (Metric) Inner Film Thickness unitSize
15278633 30 m 0.32 mm 3 μm Each
10422171 25 m 0.15 mm 1 μm Each
10107540 25 m 0.32 mm 1 μm Each
16682602 12 m 0.32 mm 0.5 μm Each
16692602 50 m 0.32 mm 0.5 μm Each
12965531 50 m 0.53 mm 5 μm Each
12653016 12 m 0.32 mm 1 μm Each
10045400 25 m 0.22 mm 0.25 μm Each
10229050 50 m 0.22 mm 0.25 μm Each
16672602 60 m 0.22 mm 0.25 μm Each
15238633 30 m 0.25 mm 1 μm Each
10310371 25 m 0.32 mm 0.25 μm Each
15268633 60 m 0.32 mm 0.5 μm Each
10441031 25 m 0.32 mm 0.5 μm Each
15298633 60 m 0.32 mm 5 μm Each
10762372 12 m 0.53 mm 1 μm Each
16602612 25 m 0.53 mm 1 μm Each
15208713 30 m 0.53 mm 5 μm Each
15218713 60 m 0.53 mm 5 μm Each
15238713 60 m 0.25 mm 0.5 μm Each
10372160 60 m 0.53 mm 3 μm Each
12796557 30 m 0.53 mm 3 μm Each
15258713 30 m 0.25 mm 0.5 μm Each
12945531 60 mm 0.25 mm 0.25 mm Each
10298760 12 mm 0.22 mm 0.25 mm Each
10177250 12 m 0.22 mm 0.1 μm Each
12975531 30 m 0.32 mm 0.25 μm Each
12654945 30 m 0.25 mm 0.25 μm Each
28 options
Product Code. 15278633
Brand: Trajan™ 054073
Only null left
Add to basket
Add to basket
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
Max. Temperature 300/320°C
Functionality/Interaction Type Van der Waals interaction
Diameter (Metric) Inner 0.32 mm
Stationary Phase 100% Dimethyl Polysiloxane
USP Type G1, G2, G38
For Use With (Application) Phenols Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines
Temperature Range -60°C to +300/320°C
Film Thickness 3 μm
Length (Metric) 30 m
Packing Material Forte™ BP1
Polarity Non polar
Coating Fused Silica