Learn More
Trajan™ BP1 GC Capillary Columns Product Code.: 15218713
  • ONLINE EXCLUSIVE PRICE
    14385.00 NOK
    14128.62 NOK / Each
    Save 256.38 NOK
    1% Off
Only null left
Add to basket
Trajan™ BP1 GC Capillary Columns
Product Code. 15218713
60 m, 0.53 mm, 5 ÎĽm, Each
Click to view available options
Length (Metric):
12 m
12 mm
25 m
30 m
50 m
60 m
60 mm
Diameter (Metric) Inner:
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 ÎĽm
0.25 ÎĽm
0.5 ÎĽm
1 ÎĽm
3 ÎĽm
5 ÎĽm
0.25 mm
Unit Size:
Each
Product Code. 15218713
Brand: Trajan™ 054807
  • ONLINE EXCLUSIVE PRICE
    14385.00 NOK
    14128.62 NOK / Each
    Save 256.38 NOK
    1% Off
Only null left
Add to basket
Only null left
Add to basket
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
Max. Temperature 280/300°C
Functionality/Interaction Type Van der Waals interaction
Diameter (Metric) Inner 0.53 mm
Stationary Phase 100% Dimethyl Polysiloxane
USP Type G1, G2, G38
For Use With (Application) Amines Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines
Temperature Range -60°C to +280/300°C
Film Thickness 5 ÎĽm
Length (Metric) 60 m
Packing Material Forte™ BP1
Polarity Non polar
Coating Fused Silica
Product Title
Trajan™ BP1 GC Capillary Columns >