Learn More
Trajan™ BP1 GC Capillary Columns
Product Code. 12975531
Change view
Click to view available options
Length (Metric):
12 m
12 mm
25 m
30 m
50 m
60 m
60 mm
Diameter (Metric) Inner:
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.5 μm
1 μm
3 μm
5 μm
0.25 mm
Unit Size:
Each
Artikelnummer. Length (Metric) Diameter (Metric) Inner Film Thickness unitSize
12975531 30 m 0.32 mm 0.25 μm Each
10422171 25 m 0.15 mm 1 μm Each
10107540 25 m 0.32 mm 1 μm Each
16682602 12 m 0.32 mm 0.5 μm Each
16692602 50 m 0.32 mm 0.5 μm Each
12965531 50 m 0.53 mm 5 μm Each
12653016 12 m 0.32 mm 1 μm Each
10045400 25 m 0.22 mm 0.25 μm Each
10229050 50 m 0.22 mm 0.25 μm Each
16672602 60 m 0.22 mm 0.25 μm Each
15238633 30 m 0.25 mm 1 μm Each
10310371 25 m 0.32 mm 0.25 μm Each
15268633 60 m 0.32 mm 0.5 μm Each
10441031 25 m 0.32 mm 0.5 μm Each
15278633 30 m 0.32 mm 3 μm Each
15298633 60 m 0.32 mm 5 μm Each
10762372 12 m 0.53 mm 1 μm Each
16602612 25 m 0.53 mm 1 μm Each
15208713 30 m 0.53 mm 5 μm Each
15218713 60 m 0.53 mm 5 μm Each
15238713 60 m 0.25 mm 0.5 μm Each
10372160 60 m 0.53 mm 3 μm Each
12796557 30 m 0.53 mm 3 μm Each
15258713 30 m 0.25 mm 0.5 μm Each
12945531 60 mm 0.25 mm 0.25 mm Each
10298760 12 mm 0.22 mm 0.25 mm Each
10177250 12 m 0.22 mm 0.1 μm Each
12654945 30 m 0.25 mm 0.25 μm Each
28 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 12975531
Marke: Trajan™ 054062
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Spezifikation
Type GC Column for Environmental Analysis
Diameter (Metric) Inner 0.32 mm
Film Thickness 0.25 μm
Length (Metric) 30 m