missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOULE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13720-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the DAZ gene family required for germ cell development. It encodes an RNA-binding protein which is more similar to Drosophila Boule than to human proteins encoded by genes DAZ (deleted in azoospermia) or DAZL (deleted in azoospermia-like). Loss of this gene function results in the absence of sperm in semen (azoospermia). Histological studies demonstrated that the primary defect is at the meiotic G2/M transition. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Specifications
BOULE | |
Polyclonal | |
Unconjugated | |
BOLL | |
bol, boule like (Drosophila), BOLL, BOULE, Protein boule like | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
501143, 66037, 75388 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N9W6, Q924M5 | |
Boll | |
BOULE Fusion Protein Ag4662 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |