Learn More
Corning™ Bottle Top Dispensers (BTD) Cylinders
Product Code. 16822100 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
2.5 mL
5 mL
10 mL
25 mL
50 mL
100 mL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
16822100 2.5 mL Each
16832100 5 mL Each
16842100 10 mL Each
16852100 25 mL Each
16862100 50 mL Each
16872100 100 mL Each
6 options
Product Code. 16822100
Brand: Corning™ 6853
Only null left
Add to basket
Add to basket
Dispenses reagent to avoid losing solution during normal transfer
  • Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
  • Comes with four adapters with different diameters for universal use with multiple bottle sizes
  • Telescopic suction tube can be adjusted smoothly to different bottle heights
  • Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
  • Every dispenser is individually tested and delivered with a certificate of performance
  • Uses:
    • For dispensing organic and inorganic liquids
  • TRUSTED_SUSTAINABILITY
    Specifications
    Volume (Metric) 2.5 mL
    For Use With (Application) Dispensing Organic and Inorganic Liquids
    Includes Four adapters