missing translation for 'onlineSavingsMsg'
Learn More

Reitenspiess™ Bottle Brush, Small

High-quality, small brush with bleached bristles and a wool head.

Brand:  Reitenspiess™ 19080334

Additional Details : Weight : 0.02000kg

Product Code. 15815342

  • 36.15 NOK / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

  • Long, thin, tube-like shape for deep reach and cleaning efficiency.
  • Bleached bristles.
  • Wool head.
  • Galvanized wire handle/stem.
Specifications

Specifications

Bottle Brush
Galvanized Wire
45 mm
Wool
150 mm
8 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers