Learn More
Reitenspiess™ Bottle Brush
Product Code. p-8008527
Change view
Click to view available options
Diameter (Metric) Bristle:
55 mm
80 mm
Bristle Material:
Bleached Bristle
Hog Bristle
Unit Size:
Each
Product Code. Diameter (Metric) Bristle Bristle Material unitSize
10196600 55 mm Hog Bristle Each
11301275 80 mm Bleached Bristle Each
2 options
Product Code. 10196600
Brand: Reitenspiess™ 22550234
Only null left
Add to basket
Edge
Add to basket
Clean, disinfect and sterilize your laboratory bottles using the Reitenspiess™ Bottle Brush. With a range of lengths and bristle types, the brush can be specified to your specific glassware needs.
  • Variety of bristle types
  • 1 each in package
  • Variety of lengths and diameters
Specifications
Product Type Bottle Brush
Bristle Material Hog Bristle
Diameter (Metric) Bristle 55 mm
Length (Metric) 500 mm
Quantity 1 Ea.