Learn More
Cole-Parmer™ Stuart™ Bottle Adapter
Product Code. 12832134
Change view
Click to view available options
Diameter (Metric) Outer:
33 mm
38 mm
40 mm
45 mm
Unit Size:
Each
Product Code. Diameter (Metric) Outer unitSize
12832134 33 mm Each
12822134 40 mm Each
12852134 45 mm Each
12842134 38 mm Each
4 options
Product Code. 12832134
Brand: Cole-Parmer™ 1009725
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Diameter (Metric) Outer 33 mm
For Use With (Equipment) BT30/50 Digital Burette
No. per Pack 1 Each
Type Buret Adapter