Learn More
VITLAB™ Borosilicate Dr. Schilling Buret
Product Code. 10300163
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10300163 25 mL Each
10679854 50 mL Each
2 options
Product Code. 10300163
Brand: VITLAB™ 106599
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Turn the burette stopcock easily to enable fine titration with the Vitlab™.Borosilicate Dr. Schilling Burette.
  • Patented VITLAB™ symbiotic (DE 10 2005 034 963) impact protection
  • Thermally stable plastic coating of the glass buret tube
  • Provides additional breaking resistance and acts as a splinter protection
Specifications
Color Brown
Capacity (Metric) 25 mL
Material Borosilicate Glass
Graduations 0.05 mL
Height (Metric) 900 mm
Includes PE-LD Reservoir Bottle
Stopcock Material PMP, PTFE
Tolerance 0.05 mL
Type Schilling Buret
Class Class B
Certifications/Compliance DIN ISO 384