Learn More
Cytiva Borosilicate Glass XK Chromatography Tubes
Product Code. 10723062 Shop All Cytiva Products
Change view
Click to view available options
Overall Length (Metric):
20 cm
30 cm
40 cm
60 cm
70 cm
100 cm
400 cm
For Use With:
16/100 Columns
16/20 Columns
16/40 Columns
16/70 Columns
26/100 Columns
26/20 Columns
26/40 Columns
26/70 Columns
50/100 Columns
50/20 Columns
50/30 Columns
50/60 Columns
Unit Size:
Each
Product Code. Overall Length (Metric) For Use With unitSize
10723062 100 cm 26/100 Columns Each
10069619 100 cm 16/100 Columns Each
10281850 20 cm 16/20 Columns Each
10567660 40 cm 16/40 Columns Each
10378659 70 cm 16/70 Columns Each
10860911 40 cm 26/40 Columns Each
10079619 70 cm 26/70 Columns Each
10753442 400 cm 50/100 Columns Each
10763442 60 cm 50/60 Columns Each
10545010 30 cm 50/30 Columns Each
10536730 20 cm 26/20 Columns Each
10516740 20 cm 50/20 Columns Each
12 options
Product Code. 10723062
Brand: Cytiva 19014701
Only null left
Add to basket
Add to basket
Can be used in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules.
  • For use with many organic solvents except chlorinated hydrocarbons, acetone and other ketones, aliphatic esters and phenol
  • Solutions containing more than 10% NaOH, 10% HCl, other strong mineral acids or 5% acetic acid should not be used
TRUSTED_SUSTAINABILITY
Specifications
For Use With 26/100 Columns
Type XK Column Tubes
Diameter (Metric) Inner 26 mm
Overall Length (Metric) 100 cm
Material Borosilicate Glass
Min. Operating Temperature (Metric) 4°C
Max. Operating Temperature (Metric) 60°C