Learn More
Kimble Chase™ Amber Vials with Caps and Liners
missing translation for 'orderingAttributeHoverText'
missing translation for 'unitSize'
Pack of 144
Description
- Amber glass vials
- Closed-top black phenolic closures with PTFE-faced white rubber liners, unattached
- Manufactured from 51 expansion borosilicate amber glass conforming to USP Type I requirements
- Alternate caps available
Specifications
Specifications
| Closure Color | Black |
| Material (Liner) | White Rubber |
| Diameter (Metric) Outer | 21 mm |
| Material | Borosilicate Glass (51 Expansion) |
| Volume (Metric) | 16 mL |
| No. per Case | 1152 |
| No. per Pack | 144 |
| Closure Material | Phenolic |
| Closure Type | Screw Cap (Closed-top) |
| Height (Metric) | 70 mm |
| Show More |