Learn More
Fisherbrand™ Borosilicate Glass Test Tube
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
75 mm
100 mm
125 mm
150 mm
Diameter (Metric):
12 mm
16 mm
24 mm
Unit Size:
Pack of 100
Pack of 50
Description
- Borosilicate glass test tubes, grade 3.3
- All ISO 4142
Specifications
Specifications
| Certifications/Compliance | ISO 4142 |
| Length (Metric) | 150 mm |
| Material | Borosilicate Glass |
| Quantity | 50 Pack |
| Diameter (Metric) | 24 mm |