Description
- Borosilicate glass test tubes, grade 3.3
- All ISO 4142
Specifications
Specifications
| Certifications/Compliance | ISO 4142 |
| Length (Metric) | 100 mm |
| Material | Borosilicate Glass |
| Quantity | 100 Pack |
| Diameter (Metric) | 12 mm |