Learn More
GPE Scientific™ Borosilicate Glass Stirrer
Description
- For use with overhead stirrer motors
- Non-whip and autoclavable
- The PTFE blades are easily removed or replaced
Specifications
Specifications
| For Use With (Equipment) | Overhead Stirrer Motors |
| Standard Taper Joint | B24 |
| Type | Uni-Form Precision Glass Stirrers |