Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug
Product Code. 10231821
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
Product Code. 10231821
Brand: Lenz Laborglasinstrumente™ 04010049
Only null left
Add to basket
Add to basket
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug are used to separate two immiscible liquids of different densities
  • With NS socket and PE stopper
  • without scale, with solid glass plug
  • with screwthread retaining device
  • material: borosilicate glass 3.3
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass
Porosity 4.0 mm
Pore Size Yes
Quantity 2Pack
Stopper Material Polyethylene