Learn More
Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug
Product Code. 10633161
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
Product Code. Capacity (Metric) unitSize
10633161 250 mL Pack of 2
10078420 1000 mL Pack of 2
10424111 2000 mL Each
10567541 100 mL Pack of 2
10415431 500 mL Pack of 2
10567351 50 mL Pack of 2
6 options
Product Code. 10633161
Brand: Lenz Laborglasinstrumente™ 04014149
Only null left
Add to basket
Add to basket
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug are manufactured from durable DURAN™ borosilicate glass.
  • Without scale
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass
Quantity 2Pack
Stopper Material Polyethylene