Learn More
Witeg™ Borosilicate Glass Soxhlet Extractor Sapparat
Product Code. 11893310
Change view
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11893310 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11893310 Supplier Witeg™ Supplier No. 2 230 500
  • ONLINE EXCLUSIVE PRICE
    4131.52 NOK
    3788.64 NOK / Pack of 1
    Save 342.88 NOK
    8% Off
 Restricted Item
Estimated Shipment: 28-12-2026
Specifications
For Use With (Equipment) Soxhlet Extraction Equipment
Product Type Soxhlet Extractor Sapparat
Capacity (Metric) 500 mL
Material Borosilicate Glass