Learn More
Kimble Chase™ Borosilicate Glass Shells Vials
Click to view available options
Volume (Metric):
2 mL
44 mL
Unit Size:
Pack of 144
Specifications
Specifications
Volume (Metric) | 2 mL |
No. per Case | 2304 |
No. per Pack | 144 |
Closure Type | Open Bottom Plug |
Documents
Scientific Resources
Catalogs
KIM001 Kimble Chase 08 09 Catalog
File size: 57.1 MB
Product Content Correction
Product Title
Kimble Chase™ Borosilicate Glass Shells Vials >