Learn More
IKA™ Borosilicate Glass RV 10.6 Reflux Distill Vertical Condensers
Shop All IKA Products
Click to view available options
Includes:
Coating
Without Coating
Unit Size:
Each
Specifications
Specifications
| Material | Borosilicate Glass |
| Type | Reflux Distill Condenser Vertical Coated |
| Includes | Coating |
| Material (Coating) | SURLYN |
| Quantity | 1 Each |