Learn More
IKA™ Borosilicate Glass RV 10.30 Vertical-Intensive condenser with Manifold
Shop All IKA Products
Click to view available options
Includes:
With Coating
Without Coating
Unit Size:
Each
Specifications
Specifications
| Material | Borosilicate Glass |
| Type | Vertical-Intensive Condenser |
| Includes | With Coating |
| Material (Coating) | SURLYN |
| Quantity | 1 Each |
| Temperature (Metric) Range | -30°C to +60°C |