Learn More
Day Impex™ Borosilicate Glass Refill
Designed with practicality and safety in mind. Day Impex™ Borosilicate Glass Refill is ultra-low evacuated to allow for safe and efficient handling of all liquified gases, freezing mixtures, and specimens.
Brand:  Day Impex™ 210017
Additional Details : Weight : 0.67200kg
 View more versions of this product
Product Code. 11807452
  • 2175.00 NOK / Each
Estimated Shipment: 02-08-2024
Description
  • Borosilicate glass inner vessels
Specifications
Specifications
138 mm
For use with Stainless Steel container.
90 mm
1 L
5 in.
80mm
Borosilicate Glass