Learn More
BRAND™ Borosilicate Glass Powder Funnel
Product Code. 10683392 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric)Top:
60 mm
70 mm
80 mm
90 mm
100 mm
120 mm
Diameter (Metric) Stem:
20 mm
22 mm
25 mm
30 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Diameter (Metric) Stem Unit Size
10683392 100 mm 25 mm Each
10468562 60 mm 20 mm Each
10623592 120 mm 30 mm Each
10568211 70 mm 20 mm Each
10049970 80 mm 22 mm Each
10181531 90 mm 25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10683392 Supplier BRAND™ Supplier No. 146519
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Manufactured from borosilicate glass. BRAND™ Borosilicate Glass Powder Funnel includes ground and fire-polished upper rim. Short, wide stem is optimal for funneling powder and other ground materials.
  • Manufactured from borosilicate glass.
  • Ground and fire-polished upper rim.
  • Short, wide stem optimal for funneling powder and other ground materials.
TRUSTED_SUSTAINABILITY
Spezifikation
Diameter (Metric)Top 100 mm
Diameter (Metric) Stem 25 mm
Material Borosilicate Glass