Learn More
BRAND™ Borosilicate Glass Powder Funnel
Product Code. 10468562 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric)Top:
60 mm
70 mm
80 mm
90 mm
100 mm
120 mm
Diameter (Metric) Stem:
18 mm
20 mm
22 mm
25 mm
30 mm
Unit Size:
Each
Product Code. Diameter (Metric)Top Diameter (Metric) Stem unitSize
10468562 60 mm 18 mm Each
10683392 100 mm 25 mm Each
10623592 120 mm 30 mm Each
10568211 70 mm 20 mm Each
10049970 80 mm 22 mm Each
10181531 90 mm 25 mm Each
6 options
Product Code. 10468562
Brand: BRAND™ 146508
Only null left
Add to basket
Add to basket
Manufactured from borosilicate glass. BRAND™ Borosilicate Glass Powder Funnel includes ground and fire-polished upper rim. Short, wide stem is optimal for funneling powder and other ground materials.
  • Manufactured from borosilicate glass.
  • Ground and fire-polished upper rim.
  • Short, wide stem optimal for funneling powder and other ground materials.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 60 mm
Diameter (Metric) Stem 18 mm
Material Borosilicate Glass