Learn More
Wheaton™ Borosilicate Glass Mortar
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
15 mL
Unit Size:
Pack of 2
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (Metric) | 2 mL |
| Material | Borosilicate Glass |
| Product Type | Mortar |
| For Use With (Equipment) | For Tissue Grinder |