Learn More
BRAND™ BISTABIL Lateral Stopcock
Shop All Brand Products
Click to view available options
Capacity (Metric):
50 mL
Unit Size:
Each
Description
- Borosilicate glass (Boro 3.3) construction
- Stopcock housing with precision Boro 3.3 burette tip
- Interchangeable borosilicate glass key with retention device
- Stopcock size 3NS/12
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| For Use With (Equipment) | Burette Cap |
| Material | Borosilicate Glass |
| No. per Pack | 1 Each |
| Includes | Retention Device |
| Type | Stopcock |